siRNA Details
| Gene Symbol: DCLK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001040260.3 | NP_001035350.2 | 11 | 2353 | ||
| NM_001040261.4 | NP_001035351.4 | 12 | 2404 | |||
| NR_036614.1 | 11 | 2350 | ||||
| XM_005262785.2 | XP_005262842.1 | 12 | 2230 | |||
| XM_005262786.2 | XP_005262843.1 | 11 | 2179 | |||
| XM_017007832.1 | XP_016863321.1 | 12 | 2233 | |||
| XM_017007833.1 | XP_016863322.1 | 10 | 1966 | |||
| GenBank mRNA | AB209181.1 | - | 11 | 2353 | ||
| AL834498.1 | - | 12 | 1451 | |||
| BC032726.1 | - | 11 | 1643 | |||

DCLK2
Genomic Map