siRNA Details
| Gene Symbol: C6orf118 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144980.3 | NP_659417.2 | 1, 2 | 57 | ||
| XM_005266838.2 | XP_005266895.1 | 1, 2 | 82 | |||
| GenBank mRNA | BC026278.1 | - | 1, 2 | 44 | 4703212 | |
| DB454755.1 | - | 1, 2 | 57 | |||

C6orf118
Genomic Map