siRNA Details
| Gene Symbol: ZNF467 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207336.1 | NP_997219.1 | 3 | 237 | ||
| XM_005249959.4 | XP_005250016.1 | 3 | 230 | |||
| XM_005249960.4 | XP_005250017.1 | 3 | 159 | |||
| XM_005249961.4 | XP_005250018.1 | 3 | 209 | |||
| XM_006715864.3 | XP_006715927.1 | 4 | 339 | |||
| XM_011515855.2 | XP_011514157.1 | 3 | 1809 | |||
| XM_011515856.2 | XP_011514158.1 | 4 | 1512 | |||
| XM_011515857.2 | XP_011514159.1 | 3 | 1810 | |||
| XM_011515858.1 | XP_011514160.1 | 4 | 424 | |||
| GenBank mRNA | BC029296.1 | - | 1 | 46 | ||
| BC052625.1 | - | 3 | 237 | 6570263 | ||

ZNF467
Genomic Map