siRNA Details
| Gene Symbol: FUT11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001284194.1 | NP_001271123.1 | 2 | 851 | ||
| NM_173540.2 | NP_775811.2 | 2 | 851 | |||
| XM_006717656.3 | XP_006717719.1 | 2 | 967 | |||
| GenBank mRNA | AK095482.1 | - | 2 | 574 | ||
| AK314944.1 | - | 2 | 833 | |||
| BC036037.1 | - | 2 | 851 | 5271548 | ||
| BC100994.1 | - | 2 | 848 | 40005868 | ||
| BC100995.1 | - | 2 | 848 | 40005869 | ||
| BC100996.1 | - | 2 | 848 | 40005870 | ||
| BC100997.1 | - | 2 | 848 | 40005873 | ||

FUT11
Genomic Map