siRNA Details
| Gene Symbol: OIT3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152635.2 | NP_689848.1 | 6, 7 | 1198 | ||
| GenBank mRNA | AK096435.1 | - | 6, 7 | 1173 | ||
| AY013707.2 | - | 6, 7 | 1029 | |||
| AY358339.1 | - | 6, 7 | 1151 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

OIT3| Gene Symbol: OIT3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152635.2 | NP_689848.1 | 6, 7 | 1198 | ||
| GenBank mRNA | AK096435.1 | - | 6, 7 | 1173 | ||
| AY013707.2 | - | 6, 7 | 1029 | |||
| AY358339.1 | - | 6, 7 | 1151 | |||