siRNA Details
| Gene Symbol: OIT3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152635.2 | NP_689848.1 | 2 | 379 | ||
| NR_130125.1 | 2 | 379 | ||||
| GenBank mRNA | AK096435.1 | - | 2 | 354 | ||
| AY013707.2 | - | 2 | 210 | |||
| AY358339.1 | - | 2 | 332 | |||
| BC126440.1 | - | 2 | 351 | 8992156 | ||
| DA979981.1 | - | 2 | 379 | |||

OIT3
Genomic Map