siRNA Details
| Gene Symbol: GSX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133267.2 | NP_573574.1 | 1 | 360 | ||
| GenBank mRNA | AB028838.1 | - | 1 | 410 | ||
| BC075089.2 | - | 1 | 118 | 30915389 | ||
| BC075090.2 | - | 1 | 118 | 30915601 | ||
| EU596451.1 | - | 1 | 360 | |||

GSX2
Genomic Map