siRNA Details
| Gene Symbol: FAM9A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001171186.1 | NP_001164657.1 | 8 | 1001 | ||
| NM_174951.3 | NP_777611.1 | 8 | 976 | |||
| GenBank mRNA | AF494343.1 | - | 8 | 976 | ||
| BC112022.1 | - | 8 | 942 | 8327490 | ||
| BC113449.1 | - | 8 | 930 | 8322501 | ||
| BC143805.1 | - | 8 | 1001 | 9052320 | ||

FAM9A
Genomic Map