siRNA Details
| Gene Symbol: ABHD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138340.4 | NP_612213.2 | 5, 6 | 808 | ||
| XM_011525847.2 | XP_011524149.1 | 5, 6 | 781 | |||
| GenBank mRNA | AF007152.1 | - | 5, 6 | 767 | ||
| AK024880.1 | - | 5, 6 | 682 | |||
| BC021196.2 | - | 5, 6 | 768 | 3942164 | ||

ABHD3
Genomic Map