siRNA Details
| Gene Symbol: PLAC4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182832.2 | NP_878252.2 | 1 | 434 | ||
| GenBank mRNA | AK092431.1 | - | 1 | 434 | ||
| BC093685.1 | - | 1 | 379 | 7939530 | ||
| BC101615.1 | - | 1 | 379 | 8069121 | ||
| BC101617.1 | - | 1 | 379 | 8069123 | ||

PLAC4
Genomic Map