siRNA Details
| Gene Symbol: KRT24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019016.2 | NP_061889.2 | 1 | 626 | ||
| XM_006721739.3 | XP_006721802.1 | 2 | 317 | |||
| XM_011524460.2 | XP_011522762.1 | 2 | 302 | |||
| XM_017024299.1 | XP_016879788.1 | 1 | 611 | |||
| GenBank mRNA | AK000268.1 | - | 1 | 626 | ||
| BC111968.1 | - | 1 | 601 | 8327436 | ||

KRT24
Genomic Map