siRNA Details
Gene Symbol: DCDC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020869.3 | NP_065920.2 | 4 | 573 | ||
GenBank mRNA | BX647348.1 | - | 4 | 573 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: DCDC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020869.3 | NP_065920.2 | 4 | 573 | ||
GenBank mRNA | BX647348.1 | - | 4 | 573 |