siRNA Details
Gene Symbol: DCDC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020869.3 | NP_065920.2 | 11 | 1636 | ||
GenBank mRNA | AB040926.1 | - | 1 | 162 | ||
BX647348.1 | - | 11 | 1636 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: DCDC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020869.3 | NP_065920.2 | 11 | 1636 | ||
GenBank mRNA | AB040926.1 | - | 1 | 162 | ||
BX647348.1 | - | 11 | 1636 |