siRNA Details
| Gene Symbol: RILPL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145058.2 | NP_659495.1 | 2 | 744 | ||
| XM_011538012.2 | XP_011536314.1 | 2 | 688 | |||
| XM_017018959.1 | XP_016874448.1 | 2 | 688 | |||
| GenBank mRNA | AB085763.1 | - | 2 | 507 | ||
| AK056934.1 | - | 2 | 728 | |||
| BC013042.2 | - | 2 | 517 | 3544761 | ||

RILPL2
Genomic Map