siRNA Details
| Gene Symbol: UBR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_174916.2 | NP_777576.1 | 36, 37 | 4125 | ||
| GenBank mRNA | AF525401.1 | - | 36, 37 | 4008 | ||
| AY061886.1 | - | 36, 37 | 4059 | |||
| BC113505.1 | - | 36, 37 | 4137 | 8322557 | ||
| BC113507.1 | - | 36, 37 | 4137 | 8322559 | ||
| BX648400.1 | - | 8, 9 | 937 | |||

UBR1
Genomic Map