siRNA Details
| Gene Symbol: ZNF778 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001201407.1 | NP_001188336.1 | 7 | 972 | ||
| NM_182531.3 | NP_872337.2 | 6 | 888 | |||
| NR_037705.1 | 6 | 845 | ||||
| XM_005256288.2 | XP_005256345.1 | 7 | 958 | |||
| XM_011522940.2 | XP_011521242.1 | 7 | 925 | |||
| XM_017023015.1 | XP_016878504.1 | 6 | 880 | |||
| XM_017023016.1 | XP_016878505.1 | 7 | 1052 | |||
| XM_017023017.1 | XP_016878506.1 | 7 | 951 | |||
| GenBank mRNA | AK056437.1 | - | 6 | 881 | ||
| BC125192.1 | - | 5 | 593 | 40122711 | ||
| BC143524.1 | - | 5 | 562 | |||

ZNF778
Genomic Map