siRNA Details
| Gene Symbol: WDR90 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145294.4 | NP_660337.3 | 37 | 4692 | ||
| XM_017023023.1 | XP_016878512.1 | 38 | 4962 | |||
| XM_017023024.1 | XP_016878513.1 | 20 | 2575 | |||
| GenBank mRNA | AK092719.1 | - | 8 | 1408 | ||
| AK093609.1 | - | 3 | 1018 | |||
| AK131510.1 | - | 20 | 2998 | |||
| BC065836.1 | - | 3 | 748 | |||

WDR90
Genomic Map