siRNA Details
| Gene Symbol: NLRC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178844.3 | NP_849172.2 | 8 | 2577 | ||
| NR_075083.2 | 8 | 2638 | ||||
| XM_017023027.1 | XP_016878516.1 | 8 | 2607 | |||
| XM_017023028.1 | XP_016878517.1 | 9 | 2781 | |||
| XM_017023029.1 | XP_016878518.1 | 10 | 2862 | |||
| XM_017023030.1 | XP_016878519.1 | 10 | 2923 | |||
| XM_017023031.1 | XP_016878520.1 | 8 | 2668 | |||
| XM_017023032.1 | XP_016878521.1 | 10 | 3624 | |||
| XM_017023033.1 | XP_016878522.1 | 10 | 2801 | |||
| XM_017023034.1 | XP_016878523.1 | 9 | 2720 | |||
| XM_017023035.1 | XP_016878524.1 | 9 | 2749 | |||
| XM_017023036.1 | XP_016878525.1 | 8 | 3308 | |||
| XM_017023037.1 | XP_016878526.1 | 9 | 2502 | |||
| XM_017023038.1 | XP_016878527.1 | 9 | 2475 | |||
| XM_017023039.1 | XP_016878528.1 | 8 | 2607 | |||
| GenBank mRNA | AF501291.1 | - | 1 | 54 | ||
| AF501292.1 | - | 1 | 54 | |||
| AK074109.1 | - | 4 | 455 | |||
| AK090431.1 | - | 8 | 2634 | |||
| AK090476.1 | - | 8 | 3692 | |||
| AY601811.1 | - | 8 | 2564 | |||
| FJ889357.1 | - | 5 | 2153 | |||

NLRC3
Genomic Map