siRNA Details
| Gene Symbol: ZNF627 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290083.1 | NP_001277012.1 | 4, 5 | 548 | ||
| NM_001290085.1 | NP_001277014.1 | 3, 4 | 389 | |||
| NM_145295.3 | NP_660338.1 | 3, 4 | 393 | |||
| XM_011527780.2 | XP_011526082.1 | 3, 4 | 281 | |||
| GenBank mRNA | AK074846.1 | - | 3, 4 | 301 | ||
| BC006279.2 | - | 3, 4 | 314 | |||
| BC069232.1 | - | 3, 4 | 278 | 6645413 | ||
| BC098416.1 | - | 3, 4 | 364 | |||
| DA786363.1 | - | 3, 4 | 349 | |||
| DB043358.1 | - | 3, 4 | 393 | |||
| DB158202.1 | - | 4, 5 | 490 | |||

ZNF627
Genomic Map