siRNA Details
| Gene Symbol: ZNF627 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290083.1 | NP_001277012.1 | 5 | 810 | ||
| NM_001290084.1 | NP_001277013.1 | 4 | 644 | |||
| NM_001290085.1 | NP_001277014.1 | 4 | 651 | |||
| NM_145295.3 | NP_660338.1 | 4 | 655 | |||
| XM_011527780.2 | XP_011526082.1 | 4 | 543 | |||
| GenBank mRNA | AF024694.1 | - | 1 | 12 | ||
| AK074846.1 | - | 4 | 563 | |||
| AK095639.1 | - | 4 | 581 | |||
| BC006279.2 | - | 4 | 576 | |||
| BC069232.1 | - | 4 | 540 | 6645413 | ||
| BC098416.1 | - | 4 | 626 | |||

ZNF627
Genomic Map