siRNA Details
| Gene Symbol: ALG14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001305242.1 | NP_001292171.1 | 1 | 162 | ||
| NM_144988.3 | NP_659425.1 | 1 | 162 | |||
| NR_131032.1 | 1 | 162 | ||||
| XM_005270582.3 | XP_005270639.1 | 1 | 184 | |||
| XM_011540897.2 | XP_011539199.1 | 1 | 184 | |||
| GenBank mRNA | AK289395.1 | - | 1 | 162 | ||
| AW242346.1 | - | 1 | 76 | |||
| BC011706.2 | - | 1 | 138 | 3689162 | ||
| HY073066.1 | - | 1 | 126 | |||

ALG14
Genomic Map