siRNA Details
| Gene Symbol: SPAG17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_206996.2 | NP_996879.1 | 14, 15 | 2052 | ||
| XM_006710426.3 | XP_006710489.1 | 14, 15 | 2052 | |||
| XM_006710427.3 | XP_006710490.1 | 13, 14 | 1857 | |||
| XM_006710429.3 | XP_006710492.1 | 1, 2 | 66 | |||
| XM_011540934.1 | XP_011539236.1 | 14, 15 | 2052 | |||
| XM_011540935.2 | XP_011539237.1 | 13, 14 | 1891 | |||
| XM_011540936.2 | XP_011539238.1 | 5, 6 | 710 | |||
| XM_011540937.2 | XP_011539239.1 | 5, 6 | 876 | |||
| GenBank mRNA | AK091816.1 | - | 14, 15 | 2038 | ||
| AY555274.1 | - | 14, 15 | 2038 | |||
| BX648110.1 | - | 14, 15 | 2053 | |||

SPAG17
Genomic Map