siRNA Details
| Gene Symbol: CRTC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181715.2 | NP_859066.1 | 14 | 2088 | ||
| XM_005244946.1 | XP_005245003.1 | 14 | 2084 | |||
| XM_005244949.2 | XP_005245006.1 | 14 | 1961 | |||
| XM_017000575.1 | XP_016856064.1 | 13 | 1857 | |||
| XM_017000576.1 | XP_016856065.1 | 13 | 1842 | |||
| XM_017000577.1 | XP_016856066.1 | 13 | 1986 | |||
| XM_017000578.1 | XP_016856067.1 | 13 | 1971 | |||
| XM_017000579.1 | XP_016856068.1 | 8 | 1383 | |||
| GenBank mRNA | AF289574.1 | - | 5 | 1047 | ||
| AK096382.1 | - | 12 | 1679 | |||
| AK223559.1 | - | 14 | 2041 | |||
| AY360172.1 | - | 14 | 2059 | |||
| AY726597.1 | - | 14 | 1868 | |||
| BC028886.1 | - | 3 | 323 | |||
| BC033304.1 | - | 13 | 1757 | |||
| BC053562.1 | - | 14 | 1999 | 6188068 | ||

CRTC2
Genomic Map