siRNA Details
| Gene Symbol: TEX37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152670.2 | NP_689883.1 | 4 | 465 | ||
| XM_005264188.3 | XP_005264245.3 | 4 | 440 | |||
| XM_011532691.1 | XP_011530993.1 | 5 | 559 | |||
| XM_011532692.1 | XP_011530994.1 | 4 | 493 | |||
| GenBank mRNA | AK058098.1 | - | 4 | 465 | ||
| AW104588.1 | - | 1 | 330 | |||
| BC029522.1 | - | 4 | 432 | 5297675 | ||

TEX37
Genomic Map