siRNA Details
| Gene Symbol: KRTCAP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001168364.1 | NP_001161836.1 | 6 | 755 | ||
| NM_001321325.1 | NP_001308254.1 | 6 | 755 | |||
| NM_173853.3 | NP_776252.2 | 6 | 755 | |||
| GenBank mRNA | AY157576.1 | - | 6 | 725 | ||
| AY358993.1 | - | 5 | 813 | |||
| BC101687.1 | - | 6 | 754 | 8069193 | ||
| BC101689.1 | - | 6 | 755 | 8069195 | ||
| BC143574.1 | - | 6 | 754 | 9052085 | ||
| CB160871.1 | - | 2 | 224 | |||

KRTCAP3
Genomic Map