siRNA Details
| Gene Symbol: KLHDC8B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173546.2 | NP_775817.1 | 2, 3 | 602 | ||
| XM_005264938.2 | XP_005264995.1 | 2, 3 | 1067 | |||
| GenBank mRNA | AK294160.1 | - | 2, 3 | 589 | ||
| AY129011.1 | - | 2, 3 | 559 | |||
| BC039083.1 | - | 2, 3 | 552 | 5167173 | ||

KLHDC8B
Genomic Map