siRNA Details
| Gene Symbol: TVP23C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC011952.1 | - | 2 | 126 | 3860433 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

TVP23C| Gene Symbol: TVP23C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC011952.1 | - | 2 | 126 | 3860433 | |