siRNA Details
| Gene Symbol: PLD6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178836.3 | NP_849158.2 | 2 | 509 | ||
| XM_017024310.1 | XP_016879799.1 | 2 | 505 | |||
| GenBank mRNA | AK090899.1 | - | 2 | 486 | ||
| AK295282.1 | - | 3 | 429 | |||
| BC031263.1 | - | 2 | 495 | 5274952 | ||
| DC326979.1 | - | 3 | 429 | |||
| Gene Symbol: FLCN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK309336.1 | - | 12 | 1279 | ||

PLD6
Genomic Map