siRNA Details
| Gene Symbol: LRRC45 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144999.3 | NP_659436.1 | 12, 13 | 1647 | ||
| GenBank mRNA | BC014109.2 | - | 12, 13 | 1545 | 4330269 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

LRRC45| Gene Symbol: LRRC45 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144999.3 | NP_659436.1 | 12, 13 | 1647 | ||
| GenBank mRNA | BC014109.2 | - | 12, 13 | 1545 | 4330269 | |