siRNA Details
| Gene Symbol: TRIM65 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173547.3 | NP_775818.2 | 5, 6 | 1047 | ||
| XM_006721760.3 | XP_006721823.1 | 5, 6 | 1084 | |||
| XM_011524499.2 | XP_011522801.1 | 5, 6 | 1087 | |||
| XM_011524500.2 | XP_011522802.1 | 5, 6 | 1088 | |||
| XM_017024343.1 | XP_016879832.1 | 4, 5 | 916 | |||
| GenBank mRNA | BC006138.1 | - | 2, 3 | 156 | ||
| BC021259.2 | - | 5, 6 | 1015 | 4128330 | ||
| BC073831.1 | - | 2, 3 | 245 | |||

TRIM65
Genomic Map