siRNA Details
| Gene Symbol: ZNF584 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001318002.1 | NP_001304931.1 | 5 | 965 | ||
| NM_173548.2 | NP_775819.1 | 4 | 867 | |||
| NR_134461.1 | 5 | 950 | ||||
| XM_005258628.4 | XP_005258685.1 | 4 | 1346 | |||
| XM_005258630.4 | XP_005258687.1 | 5 | 983 | |||
| GenBank mRNA | AK097218.1 | - | 1 | 1294 | ||
| AK290068.1 | - | 4 | 852 | |||
| AK298929.1 | - | 5 | 862 | |||
| BC017651.1 | - | 5 | 872 | |||
| BC033149.1 | - | 4 | 845 | 4125244 | ||
| DB053426.1 | - | 4 | 464 | |||

ZNF584
Genomic Map