siRNA Details
| Gene Symbol: SLC10A4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152679.3 | NP_689892.1 | 2 | 823 | ||
| GenBank mRNA | AK126542.1 | - | 2 | 823 | ||
| AY704414.1 | - | 2 | 604 | |||
| BC012048.1 | - | 2 | 681 | 3502817 | ||
| BC019066.2 | - | 2 | 734 | 4897519 | ||

SLC10A4
Genomic Map