siRNA Details
| Gene Symbol: TAPT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153365.2 | NP_699196.2 | 2 | 356 | ||
| XM_005248139.2 | XP_005248196.1 | 2 | 289 | |||
| XM_005248140.3 | XP_005248197.1 | 2 | 239 | |||
| XM_011513812.2 | XP_011512114.1 | 2 | 354 | |||
| XM_011513815.2 | XP_011512117.1 | 2 | 1574 | |||
| XM_017007876.1 | XP_016863365.1 | 2 | 180 | |||
| GenBank mRNA | AK074494.1 | - | 2 | 289 | ||
| BC066899.1 | - | 2 | 294 | 4825556 | ||
| DB461077.1 | - | 2 | 359 | |||

TAPT1
Genomic Map