siRNA Details
| Gene Symbol: TAPT1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027696.1 | 1 | 474 | |||
| NR_027697.1 | 1 | 474 | ||||
| GenBank mRNA | AK096972.1 | - | 1 | 474 | ||
| BC132927.1 | - | 1 | 474 | |||
| BF732723.1 | - | 1 | 148 | |||
| DA640966.1 | - | 1 | 97 | |||
| DR004294.1 | - | 1 | 458 | |||

TAPT1-AS1
Genomic Map