siRNA Details
| Gene Symbol: STK32A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001112724.1 | NP_001106195.1 | 4 | 480 | ||
| NM_001287740.1 | NP_001274669.1 | 4 | 480 | |||
| NM_145001.3 | NP_659438.1 | 4 | 480 | |||
| XM_011537576.1 | XP_011535878.1 | 3 | 216 | |||
| XM_011537577.2 | XP_011535879.1 | 4 | 533 | |||
| XM_011537579.2 | XP_011535881.1 | 4 | 533 | |||
| XM_017009213.1 | XP_016864702.1 | 4 | 533 | |||
| GenBank mRNA | AK094580.1 | - | 4 | 533 | ||
| AK299099.1 | - | 4 | 371 | |||
| AK315942.1 | - | 4 | 470 | |||
| BC021666.1 | - | 4 | 393 | 3682677 | ||
| DA110948.1 | - | 4 | 480 | |||
| DR006392.1 | - | 4 | 372 | |||

STK32A
Genomic Map