siRNA Details
| Gene Symbol: HTRA4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153692.3 | NP_710159.1 | 3, 4 | 884 | ||
| XM_011544431.2 | XP_011542733.1 | 3, 4 | 884 | |||
| XM_011544432.1 | XP_011542734.1 | 3, 4 | 884 | |||
| GenBank mRNA | AK075205.1 | - | 3, 4 | 855 | ||
| AK300197.1 | - | 3, 4 | 763 | |||
| BC057765.1 | - | 3, 4 | 816 | 30348811 | ||

HTRA4
Genomic Map