siRNA Details
| Gene Symbol: ERICH5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173549.2 | NP_775820.2 | 1, 2 | 211 | ||
| GenBank mRNA | AK096872.1 | - | 1, 2 | 211 | ||
| BC062359.1 | - | 1, 2 | 155 | 5015294 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ERICH5| Gene Symbol: ERICH5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173549.2 | NP_775820.2 | 1, 2 | 211 | ||
| GenBank mRNA | AK096872.1 | - | 1, 2 | 211 | ||
| BC062359.1 | - | 1, 2 | 155 | 5015294 | ||