siRNA Details
| Gene Symbol: ANO5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001142649.1 | NP_001136121.1 | 9 | 1166 | ||
| NM_213599.2 | NP_998764.1 | 9 | 1169 | |||
| XM_005252820.3 | XP_005252877.2 | 8 | 1273 | |||
| XM_005252821.3 | XP_005252878.2 | 8 | 1270 | |||
| XM_005252822.4 | XP_005252879.1 | 9 | 1054 | |||
| XM_011519949.2 | XP_011518251.1 | 7 | 1222 | |||
| GenBank mRNA | AB125267.1 | - | 9 | 1169 | ||
| AL832374.1 | - | 9 | 1191 | |||
| AL833271.1 | - | 9 | 1190 | |||

ANO5
Genomic Map