siRNA Details
| Gene Symbol: C3orf58 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134470.1 | NP_001127942.1 | 3 | 845 | ||
| NM_173552.3 | NP_775823.1 | 3 | 1799 | |||
| XM_011512543.2 | XP_011510845.1 | 2 | 1384 | |||
| XM_017005869.1 | XP_016861358.1 | 3 | 679 | |||
| GenBank mRNA | AK095063.1 | - | 1 | 164 | ||
| AK095161.1 | - | 3 | 861 | |||
| AK095400.1 | - | 3 | 705 | |||
| AK294020.1 | - | 3 | 743 | |||
| AK315082.1 | - | 3 | 1773 | |||
| AL832839.1 | - | 3 | 861 | |||
| BC037293.2 | - | 3 | 1526 | 5267770 | ||

C3orf58
Genomic Map