siRNA Details
| Gene Symbol: SENP5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001308045.1 | NP_001294974.1 | 2 | 421 | ||
| NM_152699.4 | NP_689912.2 | 2 | 421 | |||
| XM_005269310.2 | XP_005269367.1 | 2 | 421 | |||
| XM_005269312.2 | XP_005269369.1 | 2 | 421 | |||
| XM_011512544.2 | XP_011510846.1 | 3 | 466 | |||
| XM_011512545.2 | XP_011510847.1 | 2 | 421 | |||
| XM_017005870.1 | XP_016861359.1 | 3 | 421 | |||
| GenBank mRNA | AK302305.1 | - | 2 | 421 | ||
| AK313764.1 | - | 2 | 394 | |||
| BC030705.2 | - | 2 | 370 | 4824344 | ||
| DC399261.1 | - | 2 | 422 | |||

SENP5
Genomic Map