siRNA Details
| Gene Symbol: DHRSX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145177.2 | NP_660160.2 | 2 | 207 | ||
| GenBank mRNA | AJ293620.1 | - | 2 | 167 | ||
| AY358849.1 | - | 2 | 169 | |||
| BC019696.2 | - | 2 | 160 | |||
| BC032340.1 | - | 2 | 171 | 5192975 | ||
| BG703185.1 | - | 2 | 212 | |||

DHRSX
Genomic Map