siRNA Details
| Gene Symbol: USP12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182488.3 | NP_872294.2 | 4, 5 | 818 | ||
| XM_005266282.2 | XP_005266339.1 | 4, 5 | 732 | |||
| XM_011534956.2 | XP_011533258.1 | 4, 5 | 569 | |||
| XM_017020416.1 | XP_016875905.1 | 3, 4 | 489 | |||
| GenBank mRNA | AF022789.1 | - | 4, 5 | 668 | ||
| AK289685.1 | - | 4, 5 | 732 | |||
| BC026072.1 | - | 4, 5 | 819 | 4830251 | ||

USP12
Genomic Map