siRNA Details
| Gene Symbol: OR4C6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004704.1 | NP_001004704.1 | 1 | 394 | ||
| GenBank mRNA | BC137216.1 | - | 1 | 423 | 9021213 | |
| BC137218.1 | - | 1 | 423 | 9021215 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

OR4C6| Gene Symbol: OR4C6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004704.1 | NP_001004704.1 | 1 | 394 | ||
| GenBank mRNA | BC137216.1 | - | 1 | 423 | 9021213 | |
| BC137218.1 | - | 1 | 423 | 9021215 | ||