siRNA Details
| Gene Symbol: OR5D18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001952.1 | NP_001001952.1 | 1 | 265 | ||
| GenBank mRNA | BC136925.1 | - | 1 | 285 | 9020917 | |
| BC136927.1 | - | 1 | 288 | 9020919 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

OR5D18| Gene Symbol: OR5D18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001952.1 | NP_001001952.1 | 1 | 265 | ||
| GenBank mRNA | BC136925.1 | - | 1 | 285 | 9020917 | |
| BC136927.1 | - | 1 | 288 | 9020919 | ||