siRNA Details
| Gene Symbol: OR5M11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005245.1 | NP_001005245.1 | 1 | 244 | ||
| GenBank mRNA | BC136951.1 | - | 1 | 268 | 9020943 | |
| BC136953.1 | - | 1 | 268 | 9020945 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

OR5M11| Gene Symbol: OR5M11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005245.1 | NP_001005245.1 | 1 | 244 | ||
| GenBank mRNA | BC136951.1 | - | 1 | 268 | 9020943 | |
| BC136953.1 | - | 1 | 268 | 9020945 | ||