siRNA Details
| Gene Symbol: OR6T1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005187.1 | NP_001005187.1 | 1 | 868 | ||
| XM_017017325.1 | XP_016872814.1 | 2 | 783 | |||
| GenBank mRNA | BC136906.1 | - | 1 | 903 | 9020897 | |
More Information
Set Membership: |
|

OR6T1
Genomic Map