siRNA Details
Gene Symbol: OR5A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001954.1 | NP_001001954.1 | 1 | 455 | ||
GenBank mRNA | BC136991.1 | - | 1 | 478 | 9020983 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: OR5A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001954.1 | NP_001001954.1 | 1 | 455 | ||
GenBank mRNA | BC136991.1 | - | 1 | 478 | 9020983 |