siRNA Details
| Gene Symbol: OR4D6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004708.1 | NP_001004708.1 | 1 | 578 | ||
| GenBank mRNA | BC137028.1 | - | 1 | 601 | 9021024 | |
| BC137029.1 | - | 1 | 601 | 9021025 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

OR4D6| Gene Symbol: OR4D6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004708.1 | NP_001004708.1 | 1 | 578 | ||
| GenBank mRNA | BC137028.1 | - | 1 | 601 | 9021024 | |
| BC137029.1 | - | 1 | 601 | 9021025 | ||