siRNA Details
| Gene Symbol: SHANK2-AS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_073536.1 | 2 | 259 | |||
| GenBank mRNA | BC004224.2 | - | 2 | 259 | 3537945 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

SHANK2-AS3| Gene Symbol: SHANK2-AS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_073536.1 | 2 | 259 | |||
| GenBank mRNA | BC004224.2 | - | 2 | 259 | 3537945 | |