siRNA Details
Gene Symbol: SHANK2-AS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_073536.1 | 2 | 320 | |||
GenBank mRNA | BC004224.2 | - | 2 | 320 | 3537945 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: SHANK2-AS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_073536.1 | 2 | 320 | |||
GenBank mRNA | BC004224.2 | - | 2 | 320 | 3537945 |